Invitrogen
Manufacturer Code:PA577687
Catalog # PIPA577687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ENTTNTFTDFFSARD corresponding to amino acid residues 10-24 of rat Prokineticin receptor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 73; G protein-coupled receptor ZAQ; G-protein coupled receptor 73; G-protein coupled receptor ZAQ; PK-R1; Prokineticin receptor 1
Gene Aliases: EG-VEGFR1; Gpr73; Pkr1; Prokr1
UniProt ID: (Mouse) Q9JKL1, (Rat) Q8R416
Entrez Gene ID: (Mouse) 58182, (Rat) 192648
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.