R&D Systems
Manufacturer Code:IC13361G100UG
Catalog # IC13361G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 253627 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Proinsulin Phe25-Asn110 Accession # P01308 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: IDDM2 ILPR insulin IRDN MODY10 proinsulin; Insulin; Insulin A chain; Insulin B chain; preproinsulin; proinsulin
Gene Aliases: IDDM; IDDM1; IDDM2; ILPR; INS; IRDN; MODY10
UniProt ID: (Human) P01308
Entrez Gene ID: (Human) 3630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.