Invitrogen
Manufacturer Code:PA517325
Catalog # PIPA517325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Ser252 of human PHB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell receptor-associated protein 32; BAP 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; OTTHUMP00000165765 OTTHUMP00000165766 OTTHUMP00000220103 OTTHUMP00000220170 OTTHUMP00000220171; Prohibitin
Gene Aliases: Bap32; HEL-215; HEL-S-54e; PHB; PHB1
UniProt ID: (Human) P35232, (Mouse) P67778, (Rat) P67779
Entrez Gene ID: (Human) 5245, (Mouse) 18673, (Rat) 25344
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.