Proteintech
Manufacturer Code:664241IG150UL
Catalog # 50-173-6851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D9C7 |
Immunogen | Prohibitin 2 Fusion Protein Ag2977 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell associated protein; B-cell receptor associated protein 37; B-cell receptor-associated protein 37; B-cell receptor-associated protein BAP37; BAP Bap37 D prohibitin p22 PHB2 PNAS 141 prohibitin 2 REA; BAP-37; D-prohibitin; PHB2; phb2 {ECO:0000250|UniProtKB:Q99623}; Prohibitin-2; Repressor of estrogen receptor activity
Gene Aliases: AU044498; BAP; Bap-37; Bap37; Bcap27; BCAP37; hBAP; p22; PHB2; PNAS-141; REA
UniProt ID: (Human) Q99623, (Mouse) O35129, (Rat) Q5XIH7
Entrez Gene ID: (Human) 11331, (Mouse) 12034, (Rat) 114766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.