Novus Biologicals
Manufacturer Code:NB1001807
Catalog # NB1001807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Repressor of Estrogen Activity (Prohibitin 2) using the numbering given in entry NP_009204.1 (GeneID 11331). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell associated protein; B-cell receptor-associated protein BAP37; BAP Bap37 BCAP37 B-cell associated protein D-prohibitin MGC117268 p22 PNAS-141 prohibitin 2 prohibitin-2 REAB-cell receptor-associated protein BAP37 Repressor of estrogen receptor activity; D-prohibitin; PHB2; Prohibitin-2; Repressor of estrogen receptor activity
Gene Aliases: BAP; Bap37; BCAP37; hBAP; p22; PHB2; PNAS-141; REA
UniProt ID: (Human) Q99623
Entrez Gene ID: (Human) 11331
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.