Invitrogen
Manufacturer Code:MA524063
Catalog # PIMA524063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 333731 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Progranulin/PGRN Thr18-Leu589 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Acrogranin; acrogranulin; Epithelin/granulin precursor; Glycoprotein of 88 Kda; GP88; Granulin PEPI CLN11 GEP GP88 PCDGF PGRN Acrogranin Granulin-epithelin PC Cell-derived growth factor Proepithelin; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulins; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; Progranulin
Gene Aliases: epithelin; Grn; Pgrn
UniProt ID: (Mouse) P28798
Entrez Gene ID: (Mouse) 14824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.