Invitrogen
Manufacturer Code:MA1187
Catalog # PIMA1187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D4-2F1 |
Immunogen | Recombit human Progranulin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acrogranin; Epithelin precursor; Epithelin-1; Epithelin-2; Glycoprotein of 88 Kda; GP88; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin PEPI CLN11 GEP GP88 PCDGF PGRN Acrogranin Granulin-epithelin PC Cell-derived growth factor Proepithelin; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; Progranulin
Gene Aliases: CLN11; GEP; GP88; GRN; PCDGF; PEPI; PGRN
UniProt ID: (Human) P28799
Entrez Gene ID: (Human) 2896
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.