Invitrogen
Manufacturer Code:PA557338
Catalog # PIPA557338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Profilin 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D3S1319E PFL profilin II profilin-2; Profilin II; Profilin-2
Gene Aliases: D3S1319E; PFL; Pfn; PFN2
UniProt ID: (Human) P35080, (Mouse) Q9JJV2, (Rat) Q9EPC6
Entrez Gene ID: (Human) 5217, (Mouse) 18645, (Rat) 81531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.