Invitrogen
Manufacturer Code:PA517444
Catalog # PIPA517444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human profilin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin binding protein; Epididymis tissue protein Li 184a; Profilin I; profilin I profilin-1; Profilin-1
Gene Aliases: ALS18; Pfn; PFN1
UniProt ID: (Human) P07737, (Bovine) P02584, (Rat) P62963, (Mouse) P62962
Entrez Gene ID: (Human) 5216, (Bovine) 513895, (Rat) 64303, (Mouse) 18643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.