Abnova
Manufacturer Code:MAB1389
Catalog # 89-110-404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M217 |
Immunogen | Recombinant protein corresponding to N-terminus of mouse Prkcq. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant Prkcq.
For Research Use Only
Protein Aliases: A130035A12Rik; AW494342; nPKC-theta; PKC theta; PKC-0; PKC-theta; Pkcq; PKCtheta; Protein kinase C theta type
Gene Aliases: A130035A12Rik; AW494342; PKC-0; PKC-theta; Pkcq; PKCtheta; Prkcq
UniProt ID: (Mouse) Q02111
Entrez Gene ID: (Mouse) 18761
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.