Novus Biologicals
Manufacturer Code:NB10074510
Catalog # NB10074510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | APS 18 |
Immunogen | Synthetic peptide corresponding to residues C S(313) K Y N A E S T E R E S Q D T V A E N D D G(334) of humen PS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AD3EC 3.4.23.- Alzheimer disease 3 EC 3.4.23 FAD presenilin 1 Protein S182 PS-1 PS1presenilin-1 PSNL1 S182; Presenilin-1; Presenilin-1 CTF subunit; Presenilin-1 CTF12; Presenilin-1 NTF subunit; Protein S182; PS-1; PS1-CTF12
Gene Aliases: AD3; FAD; PS-1; PS1; PSEN1; PSNL1; S182
UniProt ID: (Human) P49768
Entrez Gene ID: (Human) 5663
Molecular Function:
aspartic protease
calcium-binding protein
hydrolase
membrane-bound signaling molecule
protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.