Abnova
Manufacturer Code:PAB14875
Catalog # 89-117-320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Gpcpd1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Gpcpd1.
For Research Use Only
Protein Aliases: 2310004G06Rik; 2310032D16Rik; AU015213; Glycerophosphocholine phosphodiesterase GPCPD1; Glycerophosphodiester phosphodiesterase 5; mKIAA1434; Preimplantation protein 4; putative glycerophosphocholine phosphodiesterase GPCPD1; putative glycerophosphodiester phosphodiesterase 5
Gene Aliases: 2310004G06Rik; 2310032D16Rik; AU015213; Gde5; Gpcpd1; Kiaa1434; mKIAA1434; Prei4
UniProt ID: (Mouse) Q8C0L9
Entrez Gene ID: (Mouse) 74182
Molecular Function:
hydrolase
lipase
phosphodiesterase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.