Proteintech
Manufacturer Code:661081IG150UL
Catalog # 50-173-6544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H8C4 |
Immunogen | Prealbumin/transthyretin Fusion Protein Ag19232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATTR; ATTR HsT2651 PALB Prealbumin Prealbumin/transthyretin TBPA transthyretin TTR; carpal tunnel syndrome 1; epididymis luminal protein 111; Prealbumin; prealbumin, amyloidosis type I; TBPA; thyroxine-binding prealbumin; Transthyretin
Gene Aliases: AA408768; AI787086; CTS; CTS1; D17860; HEL111; HsT2651; PALB; prealbumin; TBPA; TTR
UniProt ID: (Human) P02766, (Mouse) P07309
Entrez Gene ID: (Human) 7276, (Mouse) 22139
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.