Abnova
Manufacturer Code:MAB2327
Catalog # 89-110-934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H2-E8 |
Immunogen | Recombinant protein corresponding to amino acids 199-409 of mouse Prdm1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant Prdm1.
For Research Use Only
Protein Aliases: B lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; Blimp-1; Blimp1; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI-BF1; ZNFPR1A1
Gene Aliases: b2b1765Clo; Blimp-1; Blimp1; PRDI-BF1; Prdm1; ZNFPR1A1
UniProt ID: (Mouse) Q60636
Entrez Gene ID: (Mouse) 12142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.