Invitrogen
Manufacturer Code:PA596175
Catalog # PIPA596175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 23-230 of human PRNP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative prion protein; AltPrP; ASCR; CD230; CD230 antigen; Major prion protein; prion protein, PrP; prion protein, structural; prion-related protein; PrP; PrP27-30; PrP33-35C
Gene Aliases: AA960666; AI325101; ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; Prn; Prn-i; Prn-p; PRNP; PRP; PrP27-30; PrP33-35C; PrP<C>; PrPC; PrPSc; Sinc
UniProt ID: (Human) O60489, (Mouse) P04925, (Rat) P13852
Entrez Gene ID: (Human) 5621, (Mouse) 19122, (Rat) 24686
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.