Invitrogen
Manufacturer Code:PA579879
Catalog # PIPA579879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of mouse PRNP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD230; Major prion protein; Major prion protein PrP PrP27-30 PrP33-35C CD230 Prnp Prn-p Prp; prion protein, PrP; prion protein, structural; PrP; PrP27-30; PrP33-35C
Gene Aliases: AA960666; AI325101; CD230; Prn; Prn-i; Prn-p; Prnp; PrP; prP27-30; prP33-35C; PrP<C>; PrPC; PrPSc; Sinc
UniProt ID: (Mouse) P04925, (Rat) P13852
Entrez Gene ID: (Mouse) 19122, (Rat) 24686
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.