Invitrogen
Manufacturer Code:PA5115797
Catalog # PIPA5115797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRNP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960666 AI325101 Alternative prion protein alternative prion protein major prion protein AltPrP ASCR BSE CD230 CD230 antigen CJD GSS KURU Major prion protein major prion protein alternative prion protein major scrapie-associated fibril protein 1 MGC26679 p27-30 PK resistant core prion protein prion protein (p27-30) (Creutzfeldt-Jakob disease Gerstmann-Strausler-Scheinker syndrome fatal familial insomnia) prion protein precursor prion protein precursor PrP prion protein PrP prion protein variant a prion protein variant b prion protein PrP prion protein structural prion-related protein PRIP Prn Prn-i Prnp Prn-p PrP PrP<C> PrP27-30 PrP33-35C PrPC PrPSc Sinc SIP TSE; Alternative prion protein; AltPrP; ASCR; CD230; CD230 antigen; Major prion protein; prion protein, PrP; prion protein, structural; prion-related protein; PrP; PrP27-30; PrP33-35C
Gene Aliases: AA960666; AI325101; ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; Prn; Prn-i; Prn-p; PRNP; PRP; PrP27-30; PrP33-35C; PrP<C>; PrPC; PrPSc; Sinc
UniProt ID: (Human) O60489, (Mouse) P04925, (Rat) P13852
Entrez Gene ID: (Human) 5621, (Mouse) 19122, (Rat) 24686
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.