Invitrogen
Manufacturer Code:PA5112165
Catalog # PIPA5112165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human PrP/PRNP fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960666 AI325101 Alternative prion protein alternative prion protein major prion protein ALTPRP ASCR BOS_13027 BSE CD230 CD230 antigen CJD GSS KURU Major prion protein Major scrapie-associated fibril protein 1 MGC26679 p27-30 PK resistant core prion protein prion protein (p27-30) (Creutzfeldt-Jakob disease Gerstmann-Strausler-Scheinker syndrome fatal familial insomnia) prion protein precursor PrP prion protein PrP prion protein variant a prion protein variant b prion protein PrP prion protein structural prion-related protein PRIP Prn Prn-i PRNP Prn-p Prp PrP<C> PrP27-30 PrP33-35C PrPc PrPSc Sinc SIP TSE; Alternative prion protein; AltPrP; ASCR; CD230; CD230 antigen; Major prion protein; prion-related protein; PrP; PrP27-30; PrP33-35C
Gene Aliases: ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; PRNP; PRP; PrP27-30; PrP33-35C; PrPc
UniProt ID: (Human) O60489
Entrez Gene ID: (Human) 5621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.