Invitrogen
Manufacturer Code:PA184495
Catalog # PIPA184495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 79-97 of the N-terminus of human prion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative prion protein; AltPrP; ASCR; ASCR CD230 CJD GSS MGC26679 PRIP PrP PrP27-30 PrP33-35C PrPc PRNP; CD230; CD230 antigen; Major prion protein; Major scrapie-associated fibril protein 1; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion-related protein; PrP; PrP27-30; PrP33-35C; PrPc
Gene Aliases: ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; prn; PRNP; PRP; PrP27-30; PrP33-35C; PRPC; SIP
UniProt ID: (Bovine) F7VJQ2, (Rabbit) Q95211, (Human) O60489, (Sheep) P23907, (Chinese hamster) Q60506
Entrez Gene ID: (Bovine) 281427, (Rabbit) 100008658, (Human) 5621, (Sheep) 493887, (Chinese hamster) 100762792
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.