Invitrogen
Manufacturer Code:PA141673
Catalog # 01-675-282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 217-229 of BSE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative prion protein; AltPrP; BSE; CD230; Major prion protein; Major scrapie-associated fibril protein 1; PK resistant core; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; PrP; PrPc
Gene Aliases: AltPrP; prn; PRNP; PRP; PRPC; SIP
UniProt ID: (Bovine) F7VJQ2, (Sheep) P23907
Entrez Gene ID: (Bovine) 281427, (Sheep) 493887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.