Abnova
Manufacturer Code:PAB8471
Catalog # 89-133-687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Ppp2r3c. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ppp2r3c.
For Research Use Only
Protein Aliases: 4930511A21Rik; 5730412A08Rik; G4-1; G5pr; phosphatase subunit gene g4-1; Protein phosphatase subunit G5PR; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma
Gene Aliases: 4930511A21Rik; 5730412A08Rik; G4-1; G5pr; MNCb-1932; Ppp2r3c
UniProt ID: (Mouse) Q9JK24
Entrez Gene ID: (Mouse) 59032
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.