Abnova
Manufacturer Code:PAB18963
Catalog # 89-120-515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of mouse Pou5f1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Pou5f1.
For Research Use Only
Protein Aliases: NF-A3; Oct-3; Oct-3/4; Oct-4; Oct3; Oct3/4; Oct4; Octamer-binding protein 3; Octamer-binding protein 4; Octamer-binding transcription factor 3; OTF-3; Otf-4; Otf3; Otf3-rs7; Otf3g; Otf4; POU domain, class 5, transcription factor 1
Gene Aliases: NF-A3; Oct-3; Oct-3/4; Oct-4; Oct3; Oct3/4; Oct4; Otf-3; Otf-4; Otf3; Otf3-rs7; Otf3g; Otf4; Pou5f1
UniProt ID: (Mouse) P20263
Entrez Gene ID: (Mouse) 18999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.