Novus Biologicals
Manufacturer Code:NB200131SS
Catalog # NB018115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human protein kinase D2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APKD2 Autosomal dominant polycystic kidney disease type II protein MGC138466 MGC138468 Pc-2 PC2 PKD4 polycystic kidney disease 2 (autosomal dominant) Polycystic kidney disease 2 protein polycystin-2 Polycystwin R48321 transient receptor potential cation channel subfamily P member 2 TRPP2; Autosomal dominant polycystic kidney disease type II protein; PC2; polycystic kidney disease 2 (autosomal dominant); Polycystic kidney disease 2 protein; Polycystin-2; Polycystwin; R48321; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2
Gene Aliases: APKD2; Pc-2; PC2; PKD2; PKD4; TRPP2
UniProt ID: (Human) Q13563
Entrez Gene ID: (Human) 5311
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.