Proteintech
Manufacturer Code:222631AP150UL
Catalog # 50-173-3803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Autosomal dominant polycystic kidney disease 1 protein; Autosomal dominant polycystic kidney disease 1 protein homolog; PBP PKD1 Polycystin 1; PC1; polycystic kidney disease 1 (autosomal dominant); polycystic kidney disease 1 homolog; polycystin-1; polycystic kidney disease-associated protein; polycystin 1; Polycystin-1; transient receptor potential cation channel, subfamily P, member 1
Gene Aliases: mFLJ00285; PBP; Pc-1; PC1; PKD1; TRPP1
UniProt ID: (Human) P98161, (Mouse) O08852
Entrez Gene ID: (Human) 5310, (Mouse) 18763
Molecular Function: G-protein modulator enzyme modulator ion channel membrane-bound signaling molecule signaling molecule transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.