Abnova
Manufacturer Code:MAB3437
Catalog # 89-111-419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 18S |
Immunogen | Rat DNA Polymerase-beta protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse monoclonal antibody raised against Polb.
For Research Use Only
Protein Aliases: DNA pol beta; DNA polymerase beta; DNA-directed DNA polymerase beta; polymerase-beta
Gene Aliases: Polb
UniProt ID: (Rat) P06766
Entrez Gene ID: (Rat) 29240
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.