Invitrogen
Manufacturer Code:MA516113
Catalog # PIMA516113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Syrian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 8.1.1 |
Immunogen | Murine thymic stromal cell lines |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Syrian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: aggrus; E11; Glycoprotein 38; Gp38; OTS-8; PA2.26 antigen; Podoplanin; RP23-348F1.2 Gp38 OTS-8 RANDAM-2 T1-alpha T1a T1alpha PA2.26 antigen aggrus glycoprotein 38 transmembrane glycoprotein E11; Transmembrane glycoprotein E11
Gene Aliases: Gp38; OTS-8; Ots8; Pdpn; RANDAM-2; T1-alpha; T1a; T1alpha
UniProt ID: (Mouse) Q62011
Entrez Gene ID: (Mouse) 14726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.