R&D Systems
Manufacturer Code:AF3244
Catalog # AF3244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Podoplanin Gln21-Lys133 Accession # Q546R8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 29kDa cytosolic podoplanin intracellular domain; 36-KD aggrus Gp36 Gp38 GP40 HT1A-1 hT1alpha-1 hT1alpha-2 lung type I cell membrane associated glycoprotein lung type-I cell membrane-associated glycoprotein (T1A-2) OTS 8 OTS8 PA2.26 antigen PDPN podoplanin RANDAM-2 T1A T1A-2 T1-alpha; Aggrus; Glycoprotein 36; glycoprotein, 36-KD; Gp36; hT1alpha-1; hT1alpha-2; lung type I cell membrane associated glycoprotein; lung type-I cell membrane-associated glycoprotein (T1A-2); PA2.26 antigen; PDPN; PICD; Podoplanin; T1-alpha; T1A
Gene Aliases: AGGRUS; GP36; Gp38; GP40; HT1A-1; OTS8; PA2.26; PDPN; PSEC0003; PSEC0025; T1A; T1A-2; T1A2; TI1A
UniProt ID: (Human) Q86YL7
Entrez Gene ID: (Human) 10630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.