Proteintech
Manufacturer Code:116291AP150UL
Catalog # 50-172-7325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Podoplanin Fusion Protein Ag2201 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 29kDa cytosolic podoplanin intracellular domain; Aggrus; AGGRUS D2 40 D2 40M2A Glycoprotein 36 GP36 Gp38 GP40 HT1A 1 M2A OTS8 PA2.26 PA2.26 antigen PDPN podoplanin PSEC0025 T1 alpha T1A T1A 2; Glycoprotein 36; glycoprotein, 36-KD; Gp36; hT1alpha-1; hT1alpha-2; lung type I cell membrane associated glycoprotein; lung type-I cell membrane-associated glycoprotein (T1A-2); PA2.26 antigen; PDPN; PICD; Podoplanin; T1-alpha; T1A
Gene Aliases: AGGRUS; GP36; Gp38; GP40; HT1A-1; OTS8; PA2.26; PDPN; PSEC0003; PSEC0025; T1A; T1A-2; T1A2; TI1A
UniProt ID: (Human) Q86YL7
Entrez Gene ID: (Human) 10630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.