Novus Biologicals
Manufacturer Code:NBP245362
Catalog # NBP245362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C6 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 32-461 of human PODXL(NP_005388) produced in HEK293 cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GCTM-2 antigen; Gp200; Gp200MGC138240 PCLP1 PCLP-1 PCLPGCTM-2 antigen PCpodocalyxin podocalyxin-like Podocalyxin-like protein 1; PC; Podocalyxin; podocalyxin-like; Podocalyxin-like protein 1
Gene Aliases: Gp200; PC; PCLP; PCLP-1; PCLP1; PODXL
UniProt ID: (Human) O00592
Entrez Gene ID: (Human) 5420
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.