Proteintech
Manufacturer Code:181501AP150UL
Catalog # 50-173-2016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Podocalyxin Fusion Protein Ag12844 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCTM 2 antigen Gp200 PC PCLP PCLP 1 PCLP1 Podocalyxin podocalyxin like Podocalyxin like protein 1 PODXL RCC marker Renal Cell Carcinoma marker TRA 1 81TRA 1 60 TRA-1-60; GCTM-2 antigen; Gp200; PC; Podocalyxin; podocalyxin-like; Podocalyxin-like protein 1
Gene Aliases: AW121214; Gp200; Ly102; PC; PCLP; PCLP-1; PCLP1; PODXL
UniProt ID: (Human) O00592, (Mouse) Q9R0M4
Entrez Gene ID: (Human) 5420, (Mouse) 27205
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.