Abnova
Manufacturer Code:MAB6482
Catalog # 89-112-893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13A6 [TEC-23] |
Immunogen | BALB/c mice were immunized with pooled lipid rafts isolated from RBL-2H3 cells. Hybridoma cells were obtained after fusion of SP02 mouse myeloma cells with spleen cells of immunized mice using standard procedures. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against Plscr1.
For Research Use Only
Protein Aliases: Ca(2+)-dependent phospholipid scramblase 1; Mg(2+)-dependent nuclease; MGC93252; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1
Gene Aliases: Plscr1
UniProt ID: (Rat) P58195
Entrez Gene ID: (Rat) 117540
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.