R&D Systems
Manufacturer Code:AF5375SP
Catalog # AF5375SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin C1 Ala35-Thr950 Accession # Q9QZC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CD232; CD232 CD232 antigen plexin (semaphorin receptor) plexin C1 PLXNC1 PLXN-C1 receptor for viral semaphorin protein receptor for virally-encoded semaphorin VESPR VESPRplexin-C1 Virus-encoded semaphorin protein receptor; plexin (semaphorin receptor); Plexin-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Virus-encoded semaphorin protein receptor
Gene Aliases: CD232; PLXN-C1; PLXNC1; VESPR
UniProt ID: (Human) O60486
Entrez Gene ID: (Human) 10154
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.