Proteintech
Manufacturer Code:106021AP150UL
Catalog # 50-172-6441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Plexin B2 Fusion Protein Ag0941 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: differentially expressed in brain tumors; KIAA0315 MM1 Nbla00445 PLEXB2 Plexin B2 PLXNB2; MM1; Plexin-B2
Gene Aliases: Debt; dJ402G11.3; KIAA0315; MM1; Nbla00445; PLEXB2; plexin-B2; PLXNB2
UniProt ID: (Human) O15031, (Mouse) B2RXS4
Entrez Gene ID: (Human) 23654, (Mouse) 140570, (Rat) 315217
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.