R&D Systems
Manufacturer Code:MAB5856
Catalog # MAB5856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 707201 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Plexin A4 Thr24-Pro1237 Accession # Q9HCM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A DKFZp434G0625 FAYV2820 FLJ35026 FLJ38287 KIAA1550DKFZp566O0546 PLEXA4 plexin A4 plexin A4 B plexin-A4 PLXNA4 PLXNA4A PLXNA4B PRO34003; Plexin-A4
Gene Aliases: FAYV2820; KIAA1550; PLEXA4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2
Entrez Gene ID: (Human) 91584
Molecular Function: kinase protein kinase protein kinase receptor receptor signaling molecule transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.