R&D Systems
Manufacturer Code:MAB5486SP
Catalog # MAB5486SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 583603 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ11751 FLJ30634 KIAA0463plexin 2 OCT OCTSemaphorin receptor OCT plexin A2 PLXN2 PLXN2plexin-A2 PLXNA2 transmembrane protein OCT; plexin 2; Plexin-A2; Semaphorin receptor OCT; transmembrane protein OCT
Gene Aliases: KIAA0463; OCT; PLXN2; PLXNA2; UNQ209/PRO235
UniProt ID: (Human) O75051
Entrez Gene ID: (Human) 5362
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
signaling molecule
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.