R&D Systems
Manufacturer Code:FAB5486A
Catalog # FAB5486A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ11751 FLJ30634 KIAA0463plexin 2 OCT OCTSemaphorin receptor OCT plexin A2 PLXN2 PLXN2plexin-A2 PLXNA2 transmembrane protein OCT; plexin 2; Plexin-A2; Semaphorin receptor OCT; transmembrane protein OCT
Gene Aliases: KIAA0463; OCT; PLXN2; PLXNA2; UNQ209/PRO235
UniProt ID: (Human) O75051
Entrez Gene ID: (Human) 5362
Molecular Function: kinase protein kinase protein kinase receptor receptor signaling molecule transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.