R&D Systems
Manufacturer Code:MAB2521100
Catalog # MAB2521100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 851407 |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf</em> 21-derived recombinant human Pleiotrophin/PTN Gly33-Asp168 Accession # P21246 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
Protein Aliases: HARP HBBM HB-GAM HBGF8 HBNF HBNF1 HBNF-1 Heparin-binding brain mitogen HNGF-8 NEGF1HBGF-8 neurite growth-promoting factor 1 neurite growth-promoting factor1) OSF-1 Osteoblast-specific factor 1 pleiotrophin PTN; HB-GAM; HBBM; HBGF-8; HBNF; HBNF-1; heparin affin regulatory protein; Heparin-binding brain mitogen; Heparin-binding growth factor 8; Heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; OSF-1; Osteoblast-specific factor 1; Pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); PTN
Gene Aliases: HARP; HB-GAM; HBBM; HBGF-8; HBGF8; HBNF; HBNF-1; HBNF1; NEGF1; OSF-1; PTN
UniProt ID: (Human) P21246
Entrez Gene ID: (Human) 5764
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.