Novus Biologicals
Manufacturer Code:NBP273464B
Catalog # NBP273464B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H8 |
Immunogen | Full length recombinant protein of human Pleckstrin (NP_002655) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ27168 p47 P47Platelet 47 kDa protein pleckstrin; p47; Platelet 47 kDa protein; Pleckstrin
Gene Aliases: P47; PLEK
UniProt ID: (Human) P08567
Entrez Gene ID: (Human) 5341
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.