Invitrogen
Manufacturer Code:PA576903
Catalog # PIPA576903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Pleckstrin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p47; Platelet 47 kDa protein; Pleckstrin; Pleckstrin Platelet 47 kDa protein p47PLEK P47
Gene Aliases: 2010300B13Rik; P47; PLEK
UniProt ID: (Human) P08567, (Mouse) Q9JHK5
Entrez Gene ID: (Human) 5341, (Mouse) 56193
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.