Invitrogen
Manufacturer Code:MA120082
Catalog # PIMA120082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8F11 |
Immunogen | Purified full length native human plasminogen protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide; Angiostatin; plasmin; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen; RP1-81D8.1 DKFZp779M0222
Gene Aliases: PLG
UniProt ID: (Human) P00747
Entrez Gene ID: (Human) 5340
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.