Novus Biologicals
Manufacturer Code:NB1107926AF488
Catalog # NB1107926AG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H3 |
Immunogen | Full length native protein (purified) (Human). |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide; Angiostatin; DKFZp779M0222 EC 3.4.21 EC 3.4.21.7 plasmin plasminogen; plasmin; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen
Gene Aliases: PLG
UniProt ID: (Human) P00747
Entrez Gene ID: (Human) 5340
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.