TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay-dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay-dependent |
Western Blot (WB) | 1:500-1:2,500 |
PUBLISHED APPLICATIONS | |
---|---|
Western Blot (WB) | See 1 publications below |
Published species | Not Applicable |
Host / Isotype | Mouse / IgG1 |
Class | Monoclonal |
Type | Antibody |
Clone | 10A1 |
Immunogen | Protein purified from Human plasma |
Conjugate | Unconjugated |
Form | Liquid |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_1959775 |
A suggested positive control for this product is human plasma.
Plasminogen is a plasma protein synthesized mostly in the liver. It consists of a single polypeptide chain and has a molecular weight of 88.5 kDa. The plasma concentration is usually in the range of 70 - 200 mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Plasmin consists of a heavy (A-) chain which contains 5 kringles and a light (B-) chain with the active site. Plasmin is a trypsin-like protease whose natural substrate is fibrin, with binding sites for fibrin residing on the kringles. Plasmin is essential for fibrinolysis and decreased fibrinolytic potential due to congenital defects in plasmin can lead to recurring thrombosis, thrombophlebitis and pulmonary embolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.