Novus Biologicals
Manufacturer Code:NB300544
Catalog # NB300544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic Peptide: V(98) Y L S E C K T G I G N G Y R G T M S(116) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activation peptide; Angiostatin; DKFZp779M0222 EC 3.4.21 EC 3.4.21.7 plasmin plasminogen; plasmin; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen
Gene Aliases: PLG
UniProt ID: (Human) P00747
Entrez Gene ID: (Human) 5340
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.