Novus Biologicals
Manufacturer Code:NBP222617
Catalog # NBP222617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D10 |
Immunogen | Recombinant human KLK3(25-261aa) purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.34 Fletcher factor kallikrein B plasma (Fletcher factor) 1 kininogenin KLK3plasma kallikrein plasma kallikrein heavy chain plasma kallikrein light chain Plasma prekallikrein PPK; Fletcher factor; kallikrein B, plasma (Fletcher factor) 1; Kininogenin; PKK; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein
Gene Aliases: KLK3; KLKB1; PKK; PKKD; PPK
UniProt ID: (Human) P03952
Entrez Gene ID: (Human) 3818
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.