R&D Systems
Manufacturer Code:MAB465RSP
Catalog # MAB465RSP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 62526R |
Immunogen | <i>S. frugiperda</i> insect ovarian cell line Sf 21-derived recombinant mouse PlGF-2 Ala24-Pro158 Accession &num P49764 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: GPI11 homolog; phosphatidylinositol glycan anchor biosynthesis, class F; Phosphatidylinositol-glycan biosynthesis class F protein; PIG-F
Gene Aliases: PIGF
UniProt ID: (Human) Q07326
Entrez Gene ID: (Human) 5281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.