Abnova
Manufacturer Code:PAB5922
Catalog # 89-132-009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Pkd2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Pkd2.
For Research Use Only
Protein Aliases: C030034P18Rik; PC2; Polycystic kidney disease 2 protein homolog; polycystin 2; Polycystin-2; Transient receptor potential cation channel subfamily P member 2; TRPP2
Gene Aliases: C030034P18Rik; PC2; Pkd2; TRPP2
UniProt ID: (Mouse) O35245
Entrez Gene ID: (Mouse) 18764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.