Abnova
Manufacturer Code:PAB15798
Catalog # 89-117-861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 140 amino acids of mouse Pkd1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Pkd1.
For Research Use Only
Protein Aliases: Autosomal dominant polycystic kidney disease 1 protein homolog; FLJ00285; mFLJ00285; MGC118471; PC1; polycystic kidney disease 1 homolog; polycystin-1; polycystin 1; Polycystin-1
Gene Aliases: mFLJ00285; PC1; Pkd1
UniProt ID: (Mouse) O08852
Entrez Gene ID: (Mouse) 18763
Molecular Function:
G-protein modulator
enzyme modulator
ion channel
membrane-bound signaling molecule
signaling molecule
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.