Invitrogen
Manufacturer Code:PA529777
Catalog # PIPA529777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 269 of Human Pirin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: iron-binding nuclear protein; Pirin; pirin (iron-binding nuclear protein); pirin like; pirin probable quercetin 23-dioxygenase PIR probable quercetinase; Probable quercetin 2,3-dioxygenase PIR; Probable quercetinase
Gene Aliases: 2310042L19Rik; PIR; Pirnl
UniProt ID: (Human) O00625, (Mouse) Q9D711
Entrez Gene ID: (Human) 8544, (Mouse) 69656
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.