Invitrogen
Manufacturer Code:PA518291
Catalog # PIPA518291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GSSKKVTLSVLSREC) corresponding to the N-terminus amino acids of PIR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PIR pirin pirin (iron-binding nuclear protein); Pirin; pirin (iron-binding nuclear protein); Probable quercetin 2,3-dioxygenase PIR; Probable quercetinase
Gene Aliases: PIR
UniProt ID: (Human) O00625
Entrez Gene ID: (Human) 8544
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.