Novus Biologicals
Manufacturer Code:NBP133555
Catalog # NBP133555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Pirinin. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.13.11.24 pirin pirin (iron-binding nuclear protein) Probable quercetin 23-dioxygenase PIR Probable quercetinase; Pirin; pirin (iron-binding nuclear protein); Probable quercetin 2,3-dioxygenase PIR; Probable quercetinase
Gene Aliases: PIR
UniProt ID: (Human) O00625
Entrez Gene ID: (Human) 8544
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.